2-Chloro-5-nitrobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
OR8648-100G OR8648-500G
APOSOR8648-100GEA 102 EUR
APOSOR8648-100G APOSOR8648-500G
2-Chloro-5-nitrobenzaldehyde 97%
2-Chloro-5-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 75…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS Number: 6361-21-3
EINECS: 228-830-0