4-Bromobenzylamine hydrochloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Bromophenyl)methanamine hydrochloride
OR8654-100G OR8654-25G
APOSOR8654-100GEA 238 EUR
APOSOR8654-100G APOSOR8654-25G
4-Bromobenzylamine hydrochloride 98%
4-Bromobenzylamine hydrochloride
Formula: C₇H₉BrClN
Melting Pt: 274…276 °C
Storage Temperature: Ambient
MDL Number: MFCD00012860
CAS Number: 26177-44-6
EINECS: 247-503-3