2-(1-Pyrrolidinyl)aniline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Pyrrolidin-1-yl)aniline
OR8694-1G OR8694-250MG OR8694-5G
APOSOR8694-1GEA 94.5 EUR
APOSOR8694-1G APOSOR8694-250MG APOSOR8694-5G
2-(1-Pyrrolidinyl)aniline 97%
2-(1-Pyrrolidinyl)aniline
Formula: C₁₀H₁₄N₂
MW: 162.230 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03563756
CAS Number: 21627-58-7