2-Bromo-4-tert-butylphenol
Supplier: Apollo Scientific
Warning
OR8716-100G OR8716-25G OR8716-5G
APOSOR8716-100GEA 82.8 EUR
APOSOR8716-100G APOSOR8716-25G APOSOR8716-5G
2-Bromo-4-tert-butylphenol
2-Bromo-4-tert-butylphenol
Formula: C₁₀H₁₃BrO
MW: 229.1100 g/mol
Boiling Pt: 113 °C/9mmHg
Melting Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD02682891
CAS Number: 2198-66-5
EINECS: 218-602-9