2,4,5-Trimethoxybenzaldehyde 99%
Supplier: Apollo Scientific
Warning
OR8719-100G OR8719-25G
APOSOR8719-100GEA 58 EUR
APOSOR8719-100G APOSOR8719-25G
2,4,5-Trimethoxybenzaldehyde 99%
2,4,5-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 140 °C/4mmHg
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00003312
CAS Number: 4460-86-0
EINECS: 224-713-3