α-Bromo-4-iodotoulene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene
OR8750-100G OR8750-25G OR8750-5G
APOSOR8750-100GEA 270 EUR
APOSOR8750-100G APOSOR8750-25G APOSOR8750-5G
α-Bromo-4-iodotoulene 97%
α-Bromo-4-iodotoulene
Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 140 °C/15mmHg
Melting Pt: 78…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,II