Phenanthrene 97%
Supplier: Apollo Scientific
Warning
OR8782-100G OR8782-500G
APOSOR8782-100GEA 0 EUR
APOSOR8782-100G APOSOR8782-500G
Phenanthrene 97%
Phenanthrene
Formula: C₁₄H₁₀
Boiling Pt: 336…340 °C
Melting Pt: 95…101 °C
Density: 1.063 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9+EHS,III