ω-Bromo-3-methoxyacetophenone 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone
OR8784-25G OR8784-5G
APOSOR8784-25GEA 119 EUR
APOSOR8784-25G APOSOR8784-5G
ω-Bromo-3-methoxyacetophenone 98%
ω-Bromo-3-methoxyacetophenone
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 285
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000199
CAS Number: 5000-65-7
EINECS: 225-666-1
UN: 3261
ADR: 8 II