2-(2-(Piperazin-1-yl)ethoxy)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: 1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine
OR8790-100G OR8790-25G OR8790-500G
APOSOR8790-100GEA 72.5 EUR
APOSOR8790-100G APOSOR8790-25G APOSOR8790-500G
2-(2-(Piperazin-1-yl)ethoxy)ethanol
2-(2-(Piperazin-1-yl)ethoxy)ethanol
Formula: C₈H₁₈N₂O₂
MW: 174.2407 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074931
CAS Number: 13349-82-1