1-(2-Chloro-4-hydroxyphenyl)ethanone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2'-Chloro-4'-hydroxyacetophenone
OR8804-5G
OR8804-25G
APOSOR8804-5GEA
50.9
EUR
APOSOR8804-5G
APOSOR8804-25G
1-(2-Chloro-4-hydroxyphenyl)ethanone
1-(2-Chloro-4-hydroxyphenyl)ethanone
Formula:
C₈H₇ClO₂ Melting Pt: 110 °C Storage Temperature: Ambient |
CAS Number:
68301-59-7 |