2-Methoxy-3-pyridineboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methoxypyridine-3-boronic acid
OR8870-25G OR8870-5G OR8870-100G
APOSOR8870-25GEA 88.6 EUR
APOSOR8870-25G APOSOR8870-5G APOSOR8870-100G
2-Methoxy-3-pyridineboronic acid
2-Methoxy-3-pyridineboronic acid
Formula: C₆H₈BNO₃
MW: 152.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03411572
CAS Number: 163105-90-6