3,5-Dimethoxyphenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dimethoxybenzeneboronic acid
OR8871-1G OR8871-25G OR8871-5G
APOSOR8871-1GEA 21.9 EUR
APOSOR8871-1G APOSOR8871-25G APOSOR8871-5G
3,5-Dimethoxyphenylboronic acid
3,5-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Boiling Pt: 371…372 °C
Melting Pt: 202…207 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD03095127
CAS Number: 192182-54-0