2-Bromo-4-formylpyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-pyridinecarboxaldehyde, 2-Bromopyridine-4-carboxaldehyde
OR8883-25G OR8883-5G OR8883-100MG
APOSOR8883-25GEA 168 EUR
APOSOR8883-25G APOSOR8883-5G APOSOR8883-100MG
2-Bromo-4-formylpyridine 97%
2-Bromo-4-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD04039311
CAS Number: 118289-17-1