4-(Aminomethyl)benzonitrile 97%
Supplier: Apollo Scientific
Warning
OR8902-1G OR8902-25G OR8902-5G
APOSOR8902-1GEA 21.9 EUR
APOSOR8902-1G APOSOR8902-25G APOSOR8902-5G
4-(Aminomethyl)benzonitrile 97%
4-(Aminomethyl)benzonitrile
Formula: C₈H₈N₂
MW: 132.1625 g/mol
Boiling Pt: 160 °C/16mmHg
Melting Pt: 19…20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025578
CAS Number: 10406-25-4