3-Aminobenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Hydroxymethyl)aniline, 3-(Hydroxymethyl)aniline, 3-Aminobenzyl Alcohol, (3-Aminophenyl)methanol
OR8921-100G OR8921-25G OR8921-500G
APOSOR8921-100GEA 239 EUR
APOSOR8921-100G APOSOR8921-25G APOSOR8921-500G
3-Aminobenzyl alcohol 98%
3-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15 g/mol
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007817
CAS Number: 1877-77-6
EINECS: 217-514-8