3-Aminomethylpyridazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Pyridazin-3-ylmethanamine
OR8924-1G
OR8924-250MG
APOSOR8924-1GEA
982
EUR
APOSOR8924-1G
APOSOR8924-250MG
3-Aminomethylpyridazine
3-Aminomethylpyridazine
Formula:
C₅H₇N₃ MW: 109.13 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05664656 CAS Number: 93319-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...