5-Isoquinolinecarboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: Isoquinoline-5-carboxylic acid
OR8925-1G OR8925-250MG
APOSOR8925-1GEA 87.3 EUR
APOSOR8925-1G APOSOR8925-250MG
5-Isoquinolinecarboxylic acid
5-Isoquinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 60…62 °C/11mmHg
Melting Pt: 275…281 °C
Storage Temperature: Ambient
MDL Number: MFCD03788744
CAS Number: 27810-64-6