2-(2-Methoxyphenoxy)ethylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(2-Aminoethoxy)-2-methoxybenzene
OR8932-100G OR8932-25G
APOSOR8932-100GEA 193 EUR
APOSOR8932-100G APOSOR8932-25G
2-(2-Methoxyphenoxy)ethylamine 98%
2-(2-Methoxyphenoxy)ethylamine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 98 °C/0.4mmHg
Density: 1.11 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00235185
CAS Number: 1836-62-0
UN: 2735
ADR: 8 III