Methyl-4-aminosalicylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-amino-2-hydroxybenzoate
OR8934-100G OR8934-25G OR8934-500G
APOSOR8934-100GEA 120 EUR
APOSOR8934-100G APOSOR8934-25G APOSOR8934-500G
Methyl-4-aminosalicylate 97%
Methyl 4-aminosalicylate
Formula: C₈H₉NO₃
MW: 167.18 g/mol
Melting Pt: 124…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00088091
CAS Number: 4136-97-4
EINECS: 223-958-3