1,2-Diiodobenzene 99%
Supplier: Apollo Scientific
Danger
OR8941-100G OR8941-25G
APOSOR8941-100GEA 132 EUR
APOSOR8941-100G APOSOR8941-25G
1,2-Diiodobenzene 99%
1,2-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 282 °C/15mmHg
Melting Pt: 23.3…23.7 °C
Density: 2.524 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001038
CAS Number: 615-42-9
EINECS: 210-425-5