3-Methoxybenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Methoxybenzene-1-sulphonyl chloride
OR8943-5G OR8943-1G
APOSOR8943-5GEA 190 EUR
APOSOR8943-5G APOSOR8943-1G
3-Methoxybenzenesulphonyl chloride 95%
3-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 158…159 °C/20mmHg
Density: 1.46 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3265
ADR: 8,II