4-Bromo-2-chloro-1-iodobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-chloro-4-iodobenzene
OR8945-100G OR8945-25G OR8945-500G
APOSOR8945-100GEA 53.8 EUR
APOSOR8945-100G APOSOR8945-25G APOSOR8945-500G
4-Bromo-2-chloro-1-iodobenzene 97%
1-Bromo-3-chloro-4-iodobenzene
Formula: C₆H₃BrClI
MW: 317.35 g/mol
Boiling Pt: 160…162 °C/15mmHg
Melting Pt: 33…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00079709
CAS Number: 31928-47-9