cis-4-Hydroxy-D(+)-proline 98%
Supplier: Apollo Scientific
Warning
OR8964-100G OR8964-25G OR8964-5G
APOSOR8964-100GEA 169 EUR
APOSOR8964-100G APOSOR8964-25G APOSOR8964-5G
cis-4-Hydroxy-D(+)-proline 98%
cis-4-Hydroxy-D(+)-proline
Formula: C₅H₉NO₃
Melting Pt: 243…248 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005252
CAS Number: 2584-71-6
EINECS: 219-963-5