4-Piperidineethanol
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Hydroxyethyl)piperidine, 2-(Piperidin-4-yl)ethanol
OR8972-1G OR8972-5G
APOSOR8972-1GEA 21.9 EUR
APOSOR8972-1G APOSOR8972-5G
4-Piperidineethanol
4-Piperidineethanol
Formula: C₇H₁₅NO
Boiling Pt: 228 °C
Melting Pt: 46…50 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006008
CAS Number: 622-26-4
EINECS: 210-727-7