5-Bromo-2-chloro-3-methylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-chloro-3-picoline
OR8977-1G OR8977-25G OR8977-5G
APOSOR8977-1GEA 21.9 EUR
APOSOR8977-1G APOSOR8977-25G APOSOR8977-5G
5-Bromo-2-chloro-3-methylpyridine
5-Bromo-2-chloro-3-methylpyridine
Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 78 °C/3mmHg
Melting Pt: 40…46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095093
CAS Number: 29241-60-9