5-Bromo-3-nitropyridine-2-carbonitrile
Supplier: Apollo Scientific
Danger
Synonyms: 5-Bromo-2-cyano-3-nitropyridine, 5-Bromo-3-nitropicolinonitrile, 5-Bromo-3-nitropyridine-2-carbonitrile
OR8996-100G OR8996-25G OR8996-5G
APOSOR8996-100GEA 207 EUR
APOSOR8996-100G APOSOR8996-25G APOSOR8996-5G
5-Bromo-3-nitropyridine-2-carbonitrile
5-Bromo-3-nitropyridine-2-carbonitrile
Formula: C₆H₂BrN₃O₂
MW: 228.0030 g/mol
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD06657551
CAS Number: 573675-25-9
UN: 2811
ADR: 6.1 III