2-Thiophenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: Thiophene-2-sulphonyl chloride
OR9006-100G OR9006-25G OR9006-5G
APOSOR9006-100GEA 162 EUR
APOSOR9006-100G APOSOR9006-25G APOSOR9006-5G
2-Thiophenesulphonyl chloride 97%
2-Thiophenesulphonyl chloride
Formula: C₄H₃ClO₂S₂
Boiling Pt: 130…132 °C/14mmHg
Melting Pt: 29…32 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005426
CAS Number: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8,II