3-Bromomethylquinoxaline-2-one 97%
Supplier: Apollo Scientific
Danger
OR9013-25G OR9013-5G
APOSOR9013-25GEA 149 EUR
APOSOR9013-25G APOSOR9013-5G
3-Bromomethylquinoxaline-2-one 97%
3-Bromomethylquinoxaline-2-one
Formula: C₉H₇BrN₂O
MW: 239.07 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03412060
CAS Number: 62235-61-4
UN: 1759
ADR: 8,III