5-Chloro-2,3-dimethoxypyridine 98%
Supplier: Apollo Scientific
OR901972-10G OR901972-1G OR901972-5G
APOSOR901972-10GEA 647 EUR
APOSOR901972-10G APOSOR901972-1G APOSOR901972-5G
5-Chloro-2,3-dimethoxypyridine 98%
5-Chloro-2,3-dimethoxypyridine
Formula: C₇H₈ClNO₂
MW: 173.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972187
CAS Number: 284040-73-9