Methyl-4-hydroxynicotinate 98%
Supplier: Apollo Scientific
Synonyms: Methyl 4-hydroxynicotinate
OR901986-1G OR901986-250MG
APOSOR901986-1GEA 173 EUR
APOSOR901986-1G APOSOR901986-250MG
Methyl-4-hydroxynicotinate 98%
Methyl 4-hydroxynicotinate
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114270
CAS Number: 67367-24-2