2,2'-Dithiobis(5-nitropyridine) 98%
Supplier: Apollo Scientific
Synonyms: 1,2-Bis(5-nitropyridin-2-yl)disulphane
OR902011-1G OR902011-25G OR902011-5G
APOSOR902011-1GEA 71.2 EUR
APOSOR902011-1G APOSOR902011-25G APOSOR902011-5G
2,2'-Dithiobis(5-nitropyridine) 98%
2,2'-Dithiobis(5-nitropyridine)
Formula: C₁₀H₆N₄O₄S₂
MW: 310.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006453
CAS Number: 2127-10-8