6-Acetamido-3-chloro-2-methylpyridine 98%
Supplier: Apollo Scientific
Synonyms: 6-Acetamido-3-chloro-2-picoline
OR902130-10G OR902130-1G OR902130-5G
APOSOR902130-10GEA 502 EUR
APOSOR902130-10G APOSOR902130-1G APOSOR902130-5G
6-Acetamido-3-chloro-2-methylpyridine 98%
6-Acetamido-3-chloro-2-methylpyridine
Formula: C₈H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD08713015
CAS Number: 160115-16-2