2-Acetamido-3-methylpyridine 96%
Supplier: Apollo Scientific
Synonyms: 2-Acetamido-3-picoline, N-(3-Methylpyridin-2-yl)acetamide
OR902134-5G
APOSOR902134-5GEA 156 EUR
APOSOR902134-5G
2-Acetamido-3-methylpyridine 96%
2-Acetamido-3-methylpyridine
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00495347
CAS Number: 7463-30-1