5-Bromo-3-hydroxy-2-picoline 98%
Supplier: Apollo Scientific
Synonyms: 5-Bromo-2-methylpyridin-3-ol
OR902290-1G OR902290-250MG
APOSOR902290-1GEA 68.3 EUR
APOSOR902290-1G APOSOR902290-250MG
5-Bromo-3-hydroxy-2-picoline 98%
5-Bromo-3-hydroxy-2-picoline
Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024425
CAS Number: 91420-25-6