(4-Pyridyl)acetone 98%
Supplier: Apollo Scientific
Warning
OR9025-25G OR9025-100G OR9025-5G
APOSOR9025-25GEA 63.8 EUR
APOSOR9025-25G APOSOR9025-100G APOSOR9025-5G
(4-Pyridyl)acetone 98%
(4-Pyridyl)acetone
Formula: C₈H₉NO
MW: 135.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129043
CAS Number: 6304-16-1