5-Oxo-1-pentylpyrrolidine-3-carboxylic acid 95%
Supplier: Apollo Scientific
OR902555-10G
OR902555-1G
OR902555-5G
APOSOR902555-10GEA
647
EUR
APOSOR902555-10G
APOSOR902555-1G
APOSOR902555-5G
5-Oxo-1-pentylpyrrolidine-3-carboxylic acid 95%
5-Oxo-1-pentylpyrrolidine-3-carboxylic acid
Formula:
C₁₀H₁₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD07357567 CAS Number: 845546-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...