Nicotinamidine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: Pyridine-3-carboximidamide MonohydrochloridePyridine-3-carboximidamide hydrochloride3-Amidinopyridine hydrochloride3-Carbamimidoylpyridine Monohydrochloride3-Amidinopyridinium chloride
APOSOR9026-10GEA 265 EUR
APOSOR9026-10G APOSOR9026-1G
Nicotinamidine hydrochloride
Nicotinamidine hydrochloride
Formula: C₆H₈ClN₃
MW: 157,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054326
CAS Number: 7356-60-7