1-Isocyano-4-nitrobenzene 95%
Supplier: Apollo Scientific
Synonyms:
1-Isocyano-4-nitrobenzene, 4-Nitrophenyl Isocyanide
OR902653-100MG
OR902653-1G
OR902653-250MG
APOSOR902653-100MGEA
119
EUR
APOSOR902653-100MG
APOSOR902653-1G
APOSOR902653-250MG
1-Isocyano-4-nitrobenzene 95%
1-Isocyano-4-nitrobenzene
Formula:
C₇H₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01937394 CAS Number: 1984-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...