2-Acetyl-4-cyanophenol 98%

Supplier: Apollo Scientific

OR902766-1G OR902766-5G
APOSOR902766-1GEA 24.4 EUR
APOSOR902766-1G APOSOR902766-5G
2-Acetyl-4-cyanophenol 98%
2-Acetyl-4-cyanophenol
Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00100432
CAS Number: 35794-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR