3'-(Methylthio)acetanilide 95%

Supplier: Apollo Scientific

Synonyms: 3-Acetamidothioanisole

OR902818-25G OR902818-5G
APOSOR902818-25GEA 400 EUR
APOSOR902818-25G APOSOR902818-5G
3'-(Methylthio)acetanilide 95%
3'-(Methylthio)acetanilide
Formula: C₉H₁₁NOS
Storage Temperature: Ambient
CAS Number: 2524-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR