2,4-Dimethyl-3-hydroxypyridine 97%
Supplier: Apollo Scientific
Warning
OR9029-1G OR9029-250MG
APOSOR9029-1GEA 102 EUR
APOSOR9029-1G APOSOR9029-250MG
2,4-Dimethyl-3-hydroxypyridine 97%
2,4-Dimethyl-3-hydroxypyridine
Formula: C₇H₉NO
MW: 123.16 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD08235268
CAS Number: 27296-76-0