3,4-Xylylacetic acid 95%
Supplier: Apollo Scientific
Synonyms: 3,4-Dimethylphenylacetic acid
OR903130-5G
APOSOR903130-5GEA 59.6 EUR
APOSOR903130-5G
3,4-Xylylacetic acid 95%
3,4-Xylylacetic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664684
CAS Number: 17283-16-8