3-Ethoxy-2-nitropyridine 98%
Supplier: Apollo Scientific
OR9033-25G OR9033-5G
APOSOR9033-25GEA 0 EUR
APOSOR9033-25G APOSOR9033-5G
3-Ethoxy-2-nitropyridine 98%
3-Ethoxy-2-nitropyridine
Formula: C₇H₈N₂O₃
Boiling Pt: 156…158 °C/15mmHg
Melting Pt: 28…32 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006257
CAS Number: 74037-50-6