2,6-Dimethyl-2,5-heptadien-4-one 98%
Supplier: Apollo Scientific
Warning
OR903430-100G OR903430-25G OR903430-5G
APOSOR903430-100GEA 255 EUR
APOSOR903430-100G APOSOR903430-25G APOSOR903430-5G
2,6-Dimethyl-2,5-heptadien-4-one 98%
2,6-Dimethyl-2,5-heptadien-4-one
Formula: C₉H₁₄O
MW: 138.2069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008901
CAS Number: 504-20-1