cis-1,2-Cyclohexanedicarboxylic acid 98%
Supplier: Apollo Scientific
Synonyms: cis-Cyclohexane-1,2-dicarboxylic acid
OR903673-100G OR903673-1KG OR903673-500G
APOSOR903673-100GEA 46.4 EUR
APOSOR903673-100G APOSOR903673-1KG APOSOR903673-500G
cis-1,2-Cyclohexanedicarboxylic acid 98%
cis-1,2-Cyclohexanedicarboxylic acid
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064949
CAS Number: 610-09-3