3-Methoxy-2-nitropyridine 98%
Supplier: Apollo Scientific
Warning
OR9037-100G OR9037-25G
APOSOR9037-100GEA 33.5 EUR
APOSOR9037-100G APOSOR9037-25G
3-Methoxy-2-nitropyridine 98%
3-Methoxy-2-nitropyridine
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00222276
CAS Number: 20265-37-6