7-Chlorothiazolo[5,4-d]pyrimidine 95%
Supplier: Apollo Scientific
OR903749-1G OR903749-250MG OR903749-5G
APOSOR903749-1GEA 77 EUR
APOSOR903749-1G APOSOR903749-250MG APOSOR903749-5G
7-Chlorothiazolo[5,4-d]pyrimidine 95%
7-Chlorothiazolo[5,4-d]pyrimidine
Formula: C₅H₂ClN₃S
MW: 171.6075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16657018
CAS Number: 13316-12-6