2-Chlorothieno[2,3-d]pyrimidine 98%
Supplier: Apollo Scientific
OR903755-1G
APOSOR903755-1GEA 108 EUR
APOSOR903755-1G
2-Chlorothieno[2,3-d]pyrimidine 98%
2-Chlorothieno[2,3-d]pyrimidine
Formula: C₆H₃ClN₂S
MW: 170.6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193399
CAS Number: 83259-30-7