4-Bromo-5-methylpyrimidine 95%
Supplier: Apollo Scientific
OR903778-1G OR903778-5G OR903778-25G
APOSOR903778-1GEA 111 EUR
APOSOR903778-1G APOSOR903778-5G APOSOR903778-25G
4-Bromo-5-methylpyrimidine 95%
4-Bromo-5-methylpyrimidine
Formula: C₅H₅BrN₂
Storage Temperature: Ambient
CAS Number: 1257851-33-4