2-Methyl-6-piperazin-1-ylpyrazine 95%
Supplier: Apollo Scientific
Danger
OR9044-1G OR9044-250MG
APOSOR9044-1GEA 212 EUR
APOSOR9044-1G APOSOR9044-250MG
2-Methyl-6-piperazin-1-ylpyrazine 95%
2-Methyl-6-piperazin-1-ylpyrazine
Formula: C₉H₁₄N₄
MW: 178.24 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD08272850
CAS Number: 51047-59-7
UN: 3259
ADR: 8 III